Skip to Content
Merck
All Photos(1)

Key Documents

EZCURVCLI

Millipore

Spools clips

for use with EZ-Pak® Dispenser Curve

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€95.20

€95.20


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€95.20

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€95.20


Please contact Customer Service for Availability

Request more information

manufacturer/tradename

EZ-Pak®

Quality Level

input

sample type beer
sample type cosmetics
sample type bottled water
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Pak® Dispenser Curve

shipped in

ambient

General description

The Spools clips are an additional accessory of the EZ-Pak® Dispenser. The EZ-Pak® Dispenser Curve revolutionizes filter handling for membrane filtration method testing by combining the benefits of high-speed sterile membrane dispensing, no-touch operation, and one-handed membrane transfer. The Spools clips aid in touch-free unpacking of the membrane.

Application

The Spools clips is an additional accessory of the EZ-Pak® Dispenser Curve used in general microbiological analysis of beer, bottled Water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service