Skip to Content
Merck
All Photos(1)

Documents

EZBNZ3-160K

Millipore

Benzonase® Endonuclease ELISA Kit

detects and quantifies Benzonase nuclease in 50 μL bioprocessing samples

Synonym(s):

Endonuclease Detection Kit, Benzonase, Benzonase Endonuclease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

product name

Benzonase® Endonuclease ELISA, kit sufficient for 96 wells

biological source

Serratia marcescens

Quality Level

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (Overnight assay)

assay range

linearity: 84-111%
(94% Avarage)

recovery range: 117-130%
(124% Average)

sensitivity: 15.63 pg/mL
(LLOQ)

inter-assay cv: ≤8%
intra-assay cv: ≤8%
standard curve range: 0.015-1 ng/mL

technique(s)

ELISASandwich: suitable

input

sample type cell culture supernatant

application(s)

detection
research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The Benzonase III ELISA kit is intended for the detection of Benzonase Nuclease from Serratia marcescens. The assays is a 96 well formatt strip well ELISA, 40 samples in duplicate, detected colorimetrically. This assay is manufactured in the USA with global availability.

Research category-Process Impurity testing
Research sub-category- Endonuclease testing

Application

For the detection of Benzonase Endonuclease

Features and Benefits

  • Capture ELISA assay.
  • Highly Verified.
  • Suitable for 40 samples in Duplicate, with a 6 point Standard Crurve.

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Protocols

This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.

This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.

This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.

This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.

Related Content

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service