Skip to Content
Merck
All Photos(1)

Key Documents

EM0180

Supelco

Buffer pH 12.45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703

grade

analytical standard

Quality Level

Agency

traceable to NIST

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

Application

This pH 12.45 buffer solution is suited for use in routine analytical methods for the calibration of pH meters and maintaining the pH of solutions.

Features and Benefits

  • Traceable to NIST standards
  • A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Accompanied with a certificate of analysis (CoA) for an effective audit
  • Accurate up to +/- 0.01 at 25 °C
  • The expiry date of the product can be found in the CoA
  • Calculated uncertainty value is for the duration of transportation and storage

Specifications

Tolerance: ±0.02

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Why does the SDS Section 9 not list the pH of this material?

    1 answer
    1. This product is a pH buffer standard. The pH value is a part of the product name and clearly stated on all product information, including the SDS.

      Helpful?

  2. Hello, I did not see any warning on the product page that this item has a short shelf life. Had I known this buffer would expire in 2 months, I would have ordered at a later date. Can the people in charge of website improvements add a warning?

    1 answer
    1. This product has a tested expiration date of one year.

      The product issue you are reporting should be investigated by our Technical Service team who can assist you with further troubleshooting and/or investigation. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Report Product Issues" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service