Skip to Content
Merck
All Photos(1)

Key Documents

ELLUR

Millipore

Luminata Crescendo ELISA HRP Substrate; 200 mL

Western blotting, dot/slot blotting

Synonym(s):

ELISA HRP Substrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€305.00
200 ML
€519.00

€305.00


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
€305.00
200 ML
€519.00

About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

€305.00


Please contact Customer Service for Availability

conjugate

peroxidase conjugate

Quality Level

packaging

pkg of 200 mL

manufacturer/tradename

Luminata

assay range

sensitivity: 0.4 pg

technique(s)

ELISA: suitable

detection method

chemiluminescent

shipped in

wet ice

General description

Chemiluminescent HRP detection reagent

Application

For use on immunoblots.

Quality

The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in ELISA assays.

Storage and Stability

Stable for 1 year at 4 °C from date of shipment. Contains hydrogen peroxide. Use in well-ventilated area.

Legal Information

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service