Skip to Content
Merck
All Photos(1)

Key Documents

CP11L

Sigma-Aldrich

Anti-Integrin α₃ Mouse mAb (P1B5)

lyophilized, clone P1B5, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P1B5, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

General description

Purified mouse monoclonal antibody generated by immunizing RBF/Dn mice with the specified immunogen and fusing splenocytes with NS-1/FOX-NY mouse myeloma cells (see application references). Recognizes the α3 integrin subunit.
Recognizes the α3-subunit of the α31 integrin complex protein in human mononuclear cells.
This Anti-Integrin α₃ Mouse mAb (P1B5) is validated for use in Cell Inhibition Assays, FC, IF, IP for the detection of Integrin α₃.

Immunogen

Human
human mononuclear cells

Application

Cell Inhibition Assays (1-5 µg/ml, see application references)

Flow Cytometry (1-5 µg/ml)

Immunoblotting (not recommended)

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (1-2 µg/sample)

Paraffin Sections (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml; product will be more stable if 0.1% sodium azide is included (do not add azide if antibody is to be used with viable cells). Lyophilized antibody should be resuspended at 4°C with occasional gentle mixing for at least 2 h. Following reconstitution, refrigerate (4°C) for short-term storage (with azide) or aliquot and freeze (-20°C) for short-term storage.

Analysis Note

Positive Control
A2058 cells

Other Notes

Albelda, S.M. and Buck, C.A. 1990. FASEB J.4, 2868.
Cheresh, D.A. et al. 1990. Cell57, 59.
Krissansen, G.W. et al. 1990. J. Biol. Chem.265, 823.
Vogel, B, et al. 1990. J. Biol. Chem.265, 5934.
Wayner, E.A. et al.1988 J. Cell Biol.107, 1881.
Buck, C.A. and Horwitz, A.F. 1987. Rev. Cell Biol.3, 179.
Hynes, R.O. 1987 Cell48, 549.
Wayner, E.A. and Carter, W.G., 1987. J. Cell Biol.105, 1873.
Will partially inhibit cell-cell adhesion of E1L-8 cells. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Jia et al.
Science advances, 6(28), eaaz5894-eaaz5894 (2020-09-15)
Biologically active ligands (e.g., RGDS from fibronectin) play critical roles in the development of chemically defined biomaterials. However, recent decades have shown only limited progress in discovering novel extracellular matrix-protein-derived ligands for translational applications. Through motif analysis of evolutionarily conserved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service