Skip to Content
Merck
All Photos(1)

Key Documents

BI0349

Sigma-Aldrich

Capping Reagent B2 60% Lutidine in Acetonitrile

Biosynthesis Novabiochem®

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

color

APHA: ≤50

transition temp

flash point 10.0 °C

shipped in

ambient

storage temp.

2-30°C

Application

BI0349-0200 is packaged in 24/400 glass bottles
BI0349-0505 is packaged in GL45 glass bottles
BI0349-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (2,6-lutidine)59.0-61.0% v/vAssay (acetonitrile)39.0-41.0% v/vColor (APHA)50 maxComponent: 2,6-Lutidine Purity98.0% minComponent: Acetonitrile Purity (By GC)99.9% minFormLiquidWater (H2O)200ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service