Documents
B26524
Millipore
Analog Pressure Gauge with Sanitary Diaphragm Protection
About This Item
UNSPSC Code:
23151800
eCl@ss:
32031690
manufacturer/tradename
Pellicon®
Quality Level
fitting
3/4 in. sanitary clamp
General description
The Analog Pressure Gauge with Sanitary Diaphragm Protection is an accessory to the Pellicon® Mini Cassette Holder which has a simple design which makes process development and small scale pharmaceutical manufacturing simple and efficient.
The Pellicon® Mini holder operates one to three Pellicon® 2 Mini cassettes in parallel or Pellicon® 3 cassettes. This sanitary holder is tightened with a small torque wrench to compress the cassettes between a manifold plate that conveys fluids in and out of the cassettes and an end plate that seals the cassettes together.
The Mini holder is designed for process development and small volume pharmaceutical manufacturing.
Features and Benefits
The Mini holder and Analog Pressure Gauge with Sanitary Diaphragm Protection is designed for process development and small volume pharmaceutical manufacturing.
Legal Information
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany