Skip to Content
Merck
All Photos(1)

Key Documents

AG502-M

Sigma-Aldrich

Human IgG1 kappa

Synonym(s):

Human IgG1 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€414.00

€414.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
€414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160405
NACRES:
NA.41

€414.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

100
300

antibody form

purified immunoglobulin

manufacturer/tradename

Chemicon®

concentration

1 mg/mL (radial immunodiffusion)

target post-translational modification

unmodified

General description

Product Source: Human myeloma serum or urine.

Physical form

Format: Purified
Purified peptide.

Storage and Stability

Maintain at 2-8°C up to 6 months in undiluted aliquots. It is normal for a small amount of precipitation to occur with time, which should be removed by centrifugation.

HANDLING:

This product has been tested and found negative for HBsAG and antibodies to HIV and Hepatitis C. Safety precautions should be taken when handling potentially infective body fluids.

Analysis Note

Single arc by IEP vs. anti-whole serum, anti-human IgG. No reactivity to anti-IgG 2, 3, 4 by RID.

PURIFICATION:

Purified by ion exchange chromatography, gel filtration and protein A chromatography as appropriate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service