Skip to Content
Merck
All Photos(2)

Key Documents

ABS1488

Sigma-Aldrich

Anti-GTP cyclohydrolase 1

from rabbit, purified by affinity chromatography

Synonym(s):

EC: 3.5.4.16, GTP cyclohydrolase, GTP-CH-I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€391.00

€391.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€391.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€391.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GCH1(2643)

General description

GTP cyclohydrolase 1 (UniProt: P30793; also known as EC: 3.5.4.16, GTP cyclohydrolase, GTP-CH-I) is encoded by the GCH1 (also known as DYT5, GCH) gene (Gene ID: 2643) in human. GTP-CH-1 is a functional enzyme that is involved in step 1 of the subpathway for synthesis of 7, 8-dihydroneopterin triphosphate from GTP. It is shown to positively regulate nitric oxide synthesis in umbilical vein endothelial cells. It is expressed in epidermis and its expression is high in basal undifferentiated keratinocytes. Four isoforms of GTP-CH-1 are reported that are produced by alternative splicing. Its activity is inhibited by tetrahydrobiopterin and GTP has a positive allosteric effect on its activity. It contains three zinc binding sites (aa 141, 144, 211) and Zn2+ is shown to be essential for its catalytic activity. Its activity is also up-regulated by IFN-gamma, TNF, interleukin-1 beta, bacterial lipopolysaccharides and phenylalanine, and is down-regulated by dibutyryl-cAMP, iloprost and 8-bromo-cGMP in HUVEC. Up-regulation of its activity stimulates the production of tetrahydrobiopterin that results in elevation of endothelial nitric oxide synthase activity. GTP-CH-1 is phosphorylated by casein kinase II at serine 81 in aortic endothelial cells during oscillatory shear stress that results in increased enzyme activity. Mutations in GCH1 gene can result in hyperphenylalaninemia and depletion of dopamine and serotonin resulting in psychomotor retardation, tonicity disorders, convulsions, and abnormal movements.

Specificity

This rabbit polyclonal antibody detects GTP cyclohydrolase 1 in human cells. It targets an epitope within 15 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human GTP cyclohydrolase 1. The immunogen sequence is conserved in all four isoforms.

Application

Anti-GTP cyclohydrolase 1, Cat. No. ABS1488, is a rabbit polyclonal antibody that detectsGTP cyclohydrolase 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected GTP cyclohydrolase 1 in human vagina and human small intestine tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected GTP cyclohydrolase 1 in 10 µg of human liver tissue lysate.


Target description

~28 kDa observed; 27.90 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service