Skip to Content
Merck
All Photos(2)

Key Documents

ABN494

Sigma-Aldrich

Anti-PHOX2A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Paired-like homeobox 2A, Paired mesoderm homeobox protein 2A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€279.00

€279.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€279.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€279.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHOX2A(401)

General description

Paired mesoderm homeobox protein 2A (PHOX2A), also known as ARIX1 homeodomain protein (ARIX) and Paired-like homeobox 2A (PMX2A), along with the closely related PHOX2B, is a paired-homeodomain transcription factor that maintains the noradrenergic phenotype during embryogenesis. PHOX2 proteins help regulate endogenous hydroxylases in neural crest cells and aid in sympathetic neuron generation. PHOX2A controls only the differentiation of the main noradrenergic center of the human brain, while PHOX2B is required for the differentiation of all central and nonperipheral noradrenergic centers. PHOX2A may be involved in regulating specific expression of catecholamine biosynthetic genes. PHOX2A is linked to congenital fibrosis of extraocular muscles type 2 (CFEOM2). PHOX2 regulation may have therapeutic applications for aging or degenerative noradrenergic neuron disorders.

Immunogen

KLH-conjugated linear peptide corresponding to human PHOX2A.

Application

Anti-PHOX2A Antibody is a highly specific rabbit polyclonal antibody, that targets Phox2a & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected PHOX2A in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in rat brain tissue lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1µg/mL of this antibody detected PHOX2A in 15 µg of rat brain tissue lysate.

Target description

~32 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service