Skip to Content
Merck
All Photos(4)

Key Documents

ABD104

Sigma-Aldrich

Anti-SnoN Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ski-like protein, Ski-related oncogene, Ski-related protein, SnoN

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SKIL(6498)

General description

The Ski family of oncogenes includes Ski and Sno (Ski-related novel gene, or Ski-like). Ski family members are nuclear proteins that form homodimers and heterodimers, bind to DNA and function as transcriptional activators and repressors. The Ski proteins regulate TGFß induced gene-specific transcriptional activation by effectively repressing Smad activity and, thereby, inhibit TGFß induced cell growth and extracellular matrix production. Recently, SnoN has been shown to maintain pluripotency through selective repression of mesendodermal genes in human ES cells.

Immunogen

KLH-conjugated linear peptide corresponding to amino acids 358-372 of human SnoN protein.

Application

Immunohistochemical Analysis: A 1:50 dilution of this antibody detected SnoN in human skeletal muscle myocytes. Immunohistochemical Analysis: A 1:50 dilution of this antibody detected SnoN in human lung tissue.
Immunocytochemicaly Analysis: A 1:100 dilution of this antibody detected SnoN in H9 human embryonic stem cells.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-SnoN Antibody is validated for use in Western Blotting and Immunocytochemistry and Immunohistochemistry for the detection of SnoN.

Quality

Evaluated by Western Blot in a human placenta tissue lysate. Western Blot Analysis: 1 μg/ml of this antibody detected SnoN in a 10μg of human placenta tissue lysate.

Target description

~80 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service