Skip to Content
Merck
All Photos(2)

Key Documents

ABC2036

Sigma-Aldrich

Anti-MLKL

from rabbit

Synonym(s):

Mixed lineage kinase domain-like protein, hMLKL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€319.00

€319.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€319.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€319.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MLKL(197259)

General description

Mixed lineage kinase domain-like protein (UniProt: Q8NB16; also known as hMLKL) is encoded by the MLKL gene (Gene ID: 197259) in human. MLKL is a pseudokinase that plays a key role in TNF-induced necroptosis, a programmed cell death process. MLKL contains a protein kinase-like domain; but it lacks several residues required for activity. Its ATP-binding site is located within amino acids 209-217. Two isoforms of MLKL have been described that are produced by alternative splicing. It contains two coiled coil regions (aa 55-84 and 139-180) and coiled coil region 2 is shown to be responsible for its homotrimerization. MLKL is phosphorylated on Thr357/Ser358 by RIP3 kinase that leads to its oligomerization. The oligomeric MLKL then translocates from cytosol to plasma membrane and intracellular membranes where it bind to phosphatidylinositol and cardiolipin and disrupts membrane integrity leading to necrotic cell death. MLKL is reported to play a critical role in TNF-induced necroptosis. (Ref.: Wang, H., et al (2014). Mol. Cell 54(1); 133-146).

Specificity

This rabbit polyclonal antibody detects Mixed lineage kinase domain-like protein in human cells. It targets an epitope within the first 124 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
GST/His-tagged recombinant fragment corresponding to the first 124 amino acids from the N-terminal region of human Mixed lineage kinase domain-like protein (MLKL).

Application

Anti-MLKL, Cat. No. ABC2036, is a rabbit polyclonal antibody that detects Mixed lineage kinase domain-like protein (MLKL) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A 1:500 dilution of from a representative lot detected MLKL in 10 µg of Jurkat cell lysate.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected MLKL in human tonsil and human squamous cell carcinoma tissue sections.

Quality

Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected MLKL in 10 µg of HT-29 cell lysate.

Target description

~52 kDa observed; 54.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service