Skip to Content
Merck
All Photos(1)

Key Documents

ABC110

Sigma-Aldrich

Anti-ELMO1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Engulfment and cell motility protein 1, Protein ced-12 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ELMO1(9844)

General description

Engulfment and cell motility protein 1 (ELMO1) is a cytoplasmic protein involved in the clearance of apoptotic cells by phagocytosis. ELMO1 is activated downstream of the phosphatidylserine receptor BAI1. ELMO1 binds to DOCK180 and facilitates its transport to the plasma membrane where DOCK180 activates the GTPase RAC1, possibly via Arf6. RAC1, like other small GTPases, mediate cytoskeleton remodeling and cell motility which is necessary for the internalization of target cells.

Specificity

No homology to Rat was found.
This antibody recognizes ELMO1 at the ELMO domain.

Immunogen

Epitope: ELMO domain
KLH-conjugated linear peptide corresponding to human ELMO1 at the ELMO domain.

Application

Detect ELMO1 using this Anti-ELMO1 Antibody validated for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected ELMO1 on 10 µg of Jurkat cell lysate.

Target description

~78 kDa observed. This antibody exhibits strong affinity, high dilutions are recommended. The calculated molecular weight is 84 kDa, however ELMO1 has been shown as a ~78 kDa band in western blots (Gumienny, T.L., et al. (2001). Cell. 107:27–41.)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service