Skip to Content
Merck
All Photos(1)

Documents

ABB2-00160A

Aurora plate case of 32 plates

For use with SMC® immunoassays and the SMCxPRO® instrument

Synonym(s):

32 Plate Storage Case, Aurora Plate Case, Storage Case for Aurora Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

packaging

case of 32 plates (Aurora)

manufacturer/tradename

SMC®

technique(s)

protein quantification: suitable
single molecule counting: suitable

General description

This microplate is a superior combination of optimal clarity, thermal stability, biocompatibility, low auto-fluorescence, flatness and chemical resistance making it ideal for use in SMC® immunoassays for the SMCxPRO® instrument. Part number ABB2-00160A is a case of 32 microplates but part number 02-1008-00 contains a single plate with sealers.

Application

For use with SMC® immunoassays and the SMCxPRO® instrument

Application Note
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service