Skip to Content
Merck
All Photos(1)

Key Documents

AB9496

Sigma-Aldrich

Anti-Trace Amine Receptor 3 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€410.00

€410.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€410.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€410.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

affinity purified immunoglobulin

Quality Level

clone

polyclonal

species reactivity

human, rabbit

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

immunohistochemistry: suitable

UniProt accession no.

General description

FUNCTION: SwissProt: Q96RI9 # Orphan receptor. Could be a receptor for trace amines. Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood.
SIZE: 348 amino acids; 39016 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: Q96RI9 ## Belongs to the G-protein coupled receptor 1 family.
Synthetic peptide from the C-terminal domain of human Trace Amine Receptor 3.

Specificity

Recognizes Trace Amine Receptor 3 [TA3].

Application

Immunohistochemistry on paraffin embedded tissue sections.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Storage and Stability

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service