Skip to Content
Merck
All Photos(1)

Key Documents

AB5976

Sigma-Aldrich

Anti-Dishevelled-2 Antibody

Chemicon®, from rabbit

Synonym(s):

DVL2, DSH Homolog 2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€344.00

€344.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
€344.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€344.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DVL2(1856)

General description

DVL2 is a member of multigene family that also contains DVL1 and DVL3. The vertebrate dishevelled proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. DVL2 encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein.

Specificity

Dishevelled-2 protein (dvl-2). By Western blot the antibody shows no cross reactivity with recombinant dishevelled-1 (dvl-1) or dishevelled-3 (dvl-3) protein. Pre-adsorption of the antibody with the immunogen peptide completely abolishes its reactivity by Western blot against disheveiled-2 protein.

Immunogen

Synthetic peptide corresponding to amino acids 633-645 of human disheveiled-2 protein.

Application

Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Neural Stem Cells
This Anti-Dishevelled-2 Antibody is validated for use in WB for the detection of Dishevelled-2.
Western blot: 1:1,000 using ECL. The antibody reacts with a protein of ~95kDa.

Dilute antibody in 50 mM PBS, pH 7.4 containing 1.5% sodium chloride and 1% normal animal serum (same species as the host of the secondary antibody).

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Partially purified immunoglobulin. Liquid in PBS, pH 7.4, containing 0.01M sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots (not less than 10 μL) for up to 6 months. Avoid repeated freeze/thaw cycles. The use of high quality antiserum-grade plastic or glass is recommended.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service