Skip to Content
Merck
All Photos(1)

Key Documents

AB5048P

Sigma-Aldrich

Anti-NMDAR1 Splice Variant C2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Grin1(14810)
rat ... Grin1(24408)

Specificity

Specific for the NMDAR1 C2 splice variants. Recognizes NMDAR-1a, 1b, 2a, and 2b NR1 splice variants. {Gorter et al, (1997), "The Role of Alternative Splicing of the NMDAR1 Receptor Subunit in Synaptic Plasticity" Chapter 4, pg 99, in The Ionotropic Glutamate Receptors , Monaghan, D.T. and Weinhold, R.J. (eds), Humana Press, New Jersey) By Western blot the antibody recognizes a protein with molecular weight of 120 kDa.

Immunogen

Peptide corresponding to the C-terminus of the rat NMDAR1 C2 splice variant.

Application

This Anti-NMDAR1 Splice Variant C2 Antibody is validated for use in IH, WB for the detection of NMDAR1 Splice Variant C2.
Western blot: 1:1000 using ECL

Immunohistochemistry: 1:1,000

Optimal working dilutions must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service