Skip to Content
Merck
All Photos(1)

Key Documents

AB16941

Sigma-Aldrich

Anti-Apaf-1 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

Apoptosis Protease-activating Factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APAF1(317)

Specificity

Apoptosis is related to many diseases and is induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. The mammalian homologues of the key cell death gene CED-4 in C. elegans has been identified recently from human and mouse and was designated Apaf1 for apoptosis protease-activating factor 1(Zou et al. 1997; Cecconi et al. 1998). Apaf1 binds to cytochrome C (Apaf2) and caspase-9 (Apaf3), which leads to caspase-9 activation. Activated caspase-9 in turn cleaves and activates caspase-3 which is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis (Li et al. 1997). Apaf1 can also associate with caspase-4 and caspase-8 (Hu et al. 1998). Apaf1 is ubiquitously expressed in human tissues (Zou et al. 1997).

Immunogen

Epitope: N-terminus
Polyclonal antibody raised against a peptide corresponding to amino acids 12 to 28 of human Apaf1 (Zou et al. 1997) The sequences of the immunogenic peptide are identical between human and mouse (Zou et al. 1997; Cecconi et al. 1998).

Application

Anti-Apaf-1 Antibody, N-terminus is an antibody against Apaf-1 for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western blot: 1:1000 to 1:4000

Whole cell lysate from HeLa cells can be used as a positive control and a 130 kDa band should be detected. Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-579

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Format: Purified

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary E Lucas et al.
The Journal of biological chemistry, 284(22), 14698-14709 (2009-03-28)
Epigenetic silencing involving the aberrant DNA methylation of promoter-associated CpG islands is one mechanism leading to the inactivation of tumor suppressor genes in human cancers. However, the molecular mechanisms underlying this event remains poorly understood. TMS1/ASC is a novel proapoptotic
Carla L O'Connor et al.
Biochimica et biophysica acta, 1783(10), 1903-1913 (2008-07-02)
X-linked-inhibitor-of-apoptosis-protein (XIAP) is the most potent intracellular inhibitor of caspases-9, -3 and -7. While highly elevated XIAP levels reduce the apoptotic response to various stimuli, the potency of physiological XIAP expression to control caspase activation and the consequences of XIAP
Hui-Chen Chen et al.
Nature cell biology, 17(10), 1270-1281 (2015-09-08)
Multidomain pro-apoptotic BAX and BAK, once activated, permeabilize mitochondria to trigger apoptosis, whereas anti-apoptotic BCL-2 members preserve mitochondrial integrity. The BH3-only molecules (BH3s) promote apoptosis by either activating BAX-BAK or inactivating anti-apoptotic members. Here, we present biochemical and genetic evidence
Sheila Jeudy et al.
PloS one, 6(8), e22369-e22369 (2011-08-19)
Laminin α2 (LAMA2)-deficient congenital muscular dystrophy is a severe, early-onset disease caused by abnormal levels of laminin 211 in the basal lamina leading to muscle weakness, transient inflammation, muscle degeneration and impaired mobility. In a Lama2-deficient mouse model for this
Marco Corvaro et al.
Cancer biology & therapy, 6(2), 209-217 (2007-01-17)
The apoptosome is a multiprotein complex mediating the mitochondrial pathway of cell death. Its importance during development has been clearly demonstrated by knocking out key genes in mouse. APAF1 is the core protein of the apoptosome and its dosage is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service