Skip to Content
Merck
All Photos(1)

Key Documents

AB15067-I

Sigma-Aldrich

Anti-EMX1 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€403.00

€403.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€403.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

affinity purified immunoglobulin

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, rabbit, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

General description

Synthetic human EMX1 peptide, a.a. 241-257.

Specificity

Expected to react with both spliced isoforms of human EMX1 (UniProt Q04741).

Application

Epitope: C-terminus.

Quality

Evaluated by Western Blotting in human fetal brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected EMX1 in 10 µg of human fetal brain tissue lysate.

Physical properties

~36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Rat. Predicted to react with Mouse, Rhesus Macaque, Canine based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service