Skip to Content
Merck
All Photos(1)

Key Documents

AB1299

Sigma-Aldrich

Anti-Cortisone Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€336.00

€336.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€336.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€336.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

radioimmunoassay: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Cortisone. Crossreactivities were tested using an ELISA with cortisone-3-cmo-x-biotin: cortisol (3.4%); 11-Deoxycortisol (0.6%); 11-Dehydrocorticosterone (0.6%); Tetrahydrocortisone (0.2%); Cortisoterone, Deoxycortiscosterone, Pregesterone, Aldosterone (<0.1%); Allotetrahydrocortisol, Tetrahydrocortisol, Cortol, Cortolone, 11-Hydroxyetiochoranolone (<0.01%).

Immunogen

Cortisone-3-cmo-BSA

Application

Detect Cortisone using this Anti-Cortisone Antibody validated for use in RIA.
RIA: 1:20,000

Optimal working dilutions must be determined by end user.
Research Category
Metabolism
Research Sub Category
Metabolic Hormones & Receptors

Physical form

Rabbit serum. Liquid containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service