Skip to Content
Merck
All Photos(2)

Documents

AB110-I

Sigma-Aldrich

Anti-PNMT Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylethanolamine N-methyltransferase, PEAMT, PEMT, PEMT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PEMT(10400)

General description

PNMT, also known as Phosphatidylethanolamine N-methyltransferase, or PEMT, PEAMT, or PEMT2, and encoded by the gene PEMT/PEMPT/PNMT, catalyzes three sequential methylation reactions of phosphatidylethanolamine to produce phosphatidylcholine a major constituent of cell membranes and a primary pulmonary surfactant. PNMT/PEMT is localized to the endoplasmic reticulum membrane as well as the mitochondrial membranes. PNMT/PEMT is widely expressed, and has the highest activity in the mitochondria. The conversion of phosphatidylethanolamine to phosphatidylcholine by sequential methylation via PNMT/PEMT occurs in the liver. A different pathway for phosphatidylcholine production starting from choline also is present in cells but it does not involve PNMT/PEMT enzyme. Mutations in PNMT/PEMT protein are associated with nonalcoholic fatty liver disease (NAFLD).

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human PNMT.

Application

Anti-PNMT Antibody detects level of PNMT & has been published & validated for use in PNMT.
Immunohistochemistry Analysis: A 1:500 dilution and a 1:2,000 dilution from a representative lot detected PNMT in human adrenal gland and testis tissues, respectively.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected PNMT in 10 µg of human liver tissue lysate.

Target description

~17 kDa observed. As isoform of PNMT has been observed between 15-20 kDa (Cui, Z., et al. (1993). J Biol. Chem. 268(22):16655-16663.).

Linkage

Replaces: AB110

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service