Skip to Content
Merck
All Photos(1)

Key Documents

902451

Supelco

Stainless steel-coated PTFE tube (80 cm)

rapid-action connectors (1x coupling, 1x nipple)

Synonym(s):

Tube VA/PTFE, 80 cm, con-nip

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€2,440.00

€2,440.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 UNIT
€2,440.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

€2,440.00


Please contact Customer Service for Availability

Request a Bulk Order

shipped in

ambient

storage temp.

room temp

General description

To meet our customers’ individual installation requirements resulting from their specific operating environments, we offer tubings with different endings. The tubing range also includes a stainless steel coated PTFE-tube (length: 80 cm) with nipple (male) Type 21 and connector (female) Type 21.
with 2 rapid-action connectors (1x coupling, 1x nipple)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service