Skip to Content
Merck
All Photos(1)

Key Documents

8.43986

Sigma-Aldrich

Divinylbenzene 80%

(stabilised with 4-tert-Butylpyrocatechol) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€84.30

€84.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€84.30

About This Item

UNSPSC Code:
12352005
NACRES:
NA.22

€84.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

potency

4100 mg/kg LD50, oral (rat)
8000 mg/kg LD50, skin (rabbit)

transition temp

flash point 70 °C

density

0.915-0.921 g/cm3 at 20 °C

storage temp.

2-8°C

General description

stabilised with 4-tert-Butylpyrocatechol for synthesis

Application

Divinylbenzene 80% (stabilised with 4-tert-Butylpyrocatechol) for synthesis. CAS 1321-74-0, molar mass 130.18 g/mol.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service