Skip to Content
Merck
All Photos(1)

Documents

8.22115

Sigma-Aldrich

Dimethyl oxalate

for synthesis

Synonym(s):

Dimethyl oxalate, Oxalic acid dimethyl ester, Methyl oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O4
CAS Number:
Molecular Weight:
118.09
MDL number:
UNSPSC Code:
12352108
EC Index Number:
209-053-6

vapor pressure

3 hPa ( 20 °C)

Quality Level

form

crystals

autoignition temp.

480 °C

pH

1 (20 °C, 60 g/L in H2O)

bp

163-164 °C/1013 hPa

mp

50-53 °C

transition temp

flash point 75 °C

solubility

60 g/L

density

1.15 g/cm3 at 54 °C

storage temp.

2-30°C

InChI

1S/C4H6O4/c1-7-3(5)4(6)8-2/h1-2H3

InChI key

LOMVENUNSWAXEN-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service