Skip to Content
Merck
All Photos(1)

Documents

8.22102

Sigma-Aldrich

Isopropyl myristate

for synthesis

Synonym(s):

Isopropyl myristate, Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O2
CAS Number:
Molecular Weight:
270.45
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-751-4
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

>300 °C

potency

>10000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

140 °C/3 hPa

transition temp

flash point >150 °C

solubility

<0.05 mg/L

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

Application

  • Nanoemulsion Formulation: A study on formulating celastrol nanoemulsions using isopropyl myristate and virgin coconut oil to improve drug delivery systems (Abdullah et al., 2022).
  • Green Synthesis Approaches: Examination of green synthesis methods for producing isopropyl myristate in novel single phase mediums, contributing to sustainable chemical processes (Vadgama et al., 2015).
  • Transdermal Drug Delivery Enhancement: Investigation of the effects of isopropyl myristate as a skin penetration enhancer for improving the delivery of active pharmaceutical ingredients (Zhao et al., 2016).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.853 - 0.854
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

302.0 °F

Flash Point(C)

> 150 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service