Skip to Content
Merck
All Photos(1)

Documents

8.21710

Sigma-Aldrich

1-Bromononane

for synthesis

Synonym(s):

1-Bromononane, Nonyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8Br
CAS Number:
Molecular Weight:
207.15
MDL number:
UNSPSC Code:
12352101
EC Index Number:
211-755-2
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

201 °C/1013 hPa

transition temp

flash point 90 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3

InChI key

AYMUQTNXKPEMLM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.087 - 1.091
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service