Skip to Content
Merck
All Photos(1)

Key Documents

8.21184

Sigma-Aldrich

Trimethyl phosphate

for synthesis

Synonym(s):

Trimethyl phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€39.10
500 ML
€78.00
2.5 L
€294.00

€39.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€39.10
500 ML
€78.00
2.5 L
€294.00

About This Item

Linear Formula:
(CH3)3PO4
CAS Number:
Molecular Weight:
140.07
MDL number:
UNSPSC Code:
12352108
EC Index Number:
208-144-8
NACRES:
NA.22

€39.10


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.133 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

840 mg/kg LD50, oral (Rat)
3388 mg/kg LD50, skin (Rabbit)

bp

197 °C/1013 hPa

mp

-46 °C

transition temp

flash point >150 °C

solubility

1000 g/L

density

1.21 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OC)(OC)OC

InChI

1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3

InChI key

WVLBCYQITXONBZ-UHFFFAOYSA-N

Application


  • Manipulating the ionic conductivity and interfacial compatibility of polymer-in-dual-salt electrolytes: This study explores the role of trimethyl phosphate in enhancing the ionic conductivity and interfacial stability of polymer-in-dual-salt electrolytes, crucial for the development of extended-temperature quasi-solid metal batteries. The research highlights the compound′s effectiveness in improving battery performance across varying temperatures, a key consideration for next-generation energy storage solutions (Lin et al., 2024).

  • Concentration levels and ecological risk assessment of typical organophosphate esters in representative surface waters of a megacity: This paper assesses the environmental impact of organophosphate esters, including trimethyl phosphate, in urban water bodies. It presents data on concentration levels and discusses the ecological risks associated with these compounds, providing essential insights for environmental protection and policy-making (Qiao et al., 2024).

  • [Screening of organophosphorus flame retardant residues in rice]: Utilizing QuEChERS-gas chromatography-quadrupole time-of-flight mass spectrometry, this study identifies the presence of organophosphorus flame retardants, such as trimethyl phosphate, in rice samples. The research underscores the importance of monitoring agricultural products for chemical residues to ensure food safety (Li et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.210 - 1.215
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service