Skip to Content
Merck
All Photos(1)

Key Documents

8.14192

Sigma-Aldrich

(S)-(-)-2-Hydroxy-1,2,2-triphenylethyl acetate

for synthesis

Synonym(s):

(S)-(-)-2-Hydroxy-1,2,2-triphenylethyl acetate, HYTRA, (S)-(-)-1,1,2,-Triphenyl-1,2-ethanediole-2-acetate, (S)-(-)-Acetic acid 2-hydroxy-1,2,2-triphenylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O3
CAS Number:
Molecular Weight:
332.39
UNSPSC Code:
12352108

Quality Level

Assay

≥99.0% (HPLC)

form

crystals

mp

237-240 °C

storage temp.

2-30°C

InChI

1S/C22H20O3/c1-17(23)25-21(18-11-5-2-6-12-18)22(24,19-13-7-3-8-14-19)20-15-9-4-10-16-20/h2-16,21,24H,1H3/t21-/m0/s1

InChI key

GXLZCXZLVDUDHP-NRFANRHFSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 237 °C
Melting range (upper value): ≤ 240 °C
Spec. rotation [α²0/D (c=1 in pyridine): -216.0 - -213.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service