Skip to Content
Merck
All Photos(1)

Key Documents

810961

Sigma-Aldrich

MOD–U–CYTE VIII Media and Antisera Diluent

19-21 g/dL protein (Biuret), Cholesterol 400-500mg/dL, USA origin, Vesicular Stomatitis Virus: None detected, Bluetongue: None detected, Blood banking: suitable, media additive: suitable, Serology: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801

product name

MOD–U–CYTE VIII Media and Antisera Diluent, pack of 1 L

description

0.1% sodium azide as preservtive
Contains sodium azide less than 0.1 g/dL

Quality Level

Assay

400-500 mg/dL cholesterol basis

form

solution

shelf life

12 mo.

purified by

heat shock fractionation

packaging

pack of 1 L

origin

USA origin

concentration

19-21 g/dL protein (Biuret)

technique(s)

blood typing: suitable

dilution

(Immunohematology)
(Serology)
(media additive)

impurities

Tested for IgG

pH

7.2-7.4

foreign activity

Bluetongue, none detected
Microbial Content tested by USP
Vesicular Stomatitis Virus, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service