Skip to Content
Merck
All Photos(1)

Key Documents

8.01847

Sigma-Aldrich

n-Butyl vinyl ether

(stabilised with 100 ppm KOH) for synthesis

Synonym(s):

n-Butyl vinyl ether, Vinyl butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-860-7

vapor pressure

51 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

190 °C (DIN 51794)

potency

10000 mg/kg LD50, oral (Rat)
3300 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-8.6 % (v/v)

bp

93-94 °C/1013 hPa

mp

-113 °C

transition temp

flash point -6 °C (DIN 51755 Part 1)

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCC)C=C

InChI

1S/C6H12O/c1-3-5-6-7-4-2/h4H,2-3,5-6H2,1H3

InChI key

UZKWTJUDCOPSNM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.778 - 0.779
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service