Skip to Content
Merck
All Photos(1)

Key Documents

71242

Sigma-Aldrich

pIEx-5 DNA - Novagen®

Novagen′s pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera derived insect cells.

Synonym(s):

InsectDirect System- pIEx-5 DNA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€472.00

€472.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€472.00

About This Item

UNSPSC Code:
41106611
NACRES:
NA.51

€472.00


Please contact Customer Service for Availability

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1.

pIEx-5 encodes the signal peptide of adipokinetic hormone (AKH) to allow secretion of the expressed protein, and C-terminal S•Tag and His•Tag coding sequences. For sequencing or PCR of recombinants in pIEx-5, refer to the table below to choose the appropriate primers.

Warning

Toxicity: Standard Handling (A)

Legal Information

INSECTDIRECT is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service