Skip to Content
Merck
All Photos(1)

Documents

681679-M

Millipore

Iron Chelator IV, 21H7

The Iron Chelator IV, 21H7 controls the biological activity of iron-regulated enzymes. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Iron Chelator IV, 21H7, 6-Bromo-Nʹ-(2-hydroxybenzylidene)-2-methylquinoline-4-carbohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14BrN3O2
Molecular Weight:
384.23
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL, light yellow

storage temp.

2-8°C

General description

A cell-permeable salicylaldehyde-acylhydrazone iron chelator that is 20-times more efficient than DFO/desferrioxamine (Cat. No. 252750) in depleting intracellular iron in colon cancer SW480 cells. Iron chelators exert their biological activities by affecting iron-regulated enzymes and singaling events, including HIF1α transcription activation (Effective [21H7] = 10 µM in SW480 and DLD-1 cells) due to inhibition of PHD- (prolyl hydroxylase) mediated HIF1α degradation, as well as stalling and enhancing, respectively, Ferritin and TfRI (Transferrin Receptor I) mRNA translation (Effective [21H7] = 10 µM in SW480 cells) due to iron depletion-induced IRP (Iron Regulatory Protein) IREs (Iron Response Elements) binding. Iron depletion is also reported to inhibit the growths of colorectal adenocarcinoma cultures, DLD-1 (IC50 = 0.6 and 2.9 µM, respectively, by 21H7 and DFO) and SW480 (IC50 = 1.0 and 3.8 µM, respectively, by 21H7 and DFO), as a result of Wnt signaling pathway blockage (Effective conc. = 5 µM 21H7 or 100 µM DFO).
A cell-permeable salicylaldehyde-acylhydrazone iron chelator that is 20-times more efficient than DFO/desferrioxamine (Cat. No. 252750) in depleting intracellular iron in colon cancer SW480 cells. Iron chelators exert their biological activities by affecting iron-regulated enzymes and singaling events, including HIF1α transcription activation due to inhibition of PHD- (prolyl hydroxylase) mediated HIF1α degradation, as well as altered mRNA translations due to enhanced IRP (Iron Regulatory Protein) IREs (Iron Response Elements) binding. Cellular iron depletion is also reported to inhibit the growths of colorectal adenocarcinoma cultures, DLD-1 (IC50 = 0.6 and 2.9 µM, respectively, by 21H7 and DFO) and SW480 (IC50 = 1.0 and 3.8 µM, respectively, by 21H7 and DFO), as a result of Wnt signaling pathway blockage (Effective conc. = 5 µM 21H7 or 100 µM DFO).

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming (45°C) is required for complete solubilization.

Other Notes

Song, S., et al. 2011. Cancer Res.71, 7628.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service