Skip to Content
Merck
All Photos(1)

Key Documents

5.31976

Sigma-Aldrich

MPS1 Inhibitor III, AZ3146

Synonym(s):

MPS1 Inhibitor III, AZ3146, AZ 3146, AZ-3146, Monopolar Spindle 1 Kinase Inhibitor, AZ3146

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€180.00

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€180.00

About This Item

Empirical Formula (Hill Notation):
C24H32N6O3
CAS Number:
Molecular Weight:
452.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

~35 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

purple

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

N1(CCC(CC1)Oc2cc(c(cc2)Nc3nc4[n]([c]([n](c4cn3)C)=O)C5CCCC5)OC)C

InChI

1S/C24H32N6O3/c1-28-12-10-17(11-13-28)33-18-8-9-19(21(14-18)32-3)26-23-25-15-20-22(27-23)30(24(31)29(20)2)16-6-4-5-7-16/h8-9,14-17H,4-7,10-13H2,1-3H3,(H,25,26,27)

General description

A cell permeable purinone derivative that acts as a highly potent and selective inhibitor of monopolar spindle 1 kinase (MPS1; IC50 = ~35 nM). Inhibits the phosphorylation of full-length immunoprecipitated human MPS1 and exhibits minimal activity against a panel of 49 other protein kinases. Has much reduced activity against FAK, JNK1, JNK2, and KIT and does not affect the activity of mitotic-specific phosphoforms of Aurora B and BubR1. Induces a significant reduction in time to complete mitosis (from 90 min to 32 min) and causes abnormal mitosis where over 50% of cells enter anaphase without aligning their chromosomes.
A cell permeable purinone derivative that acts as a highly potent and selective inhibitor of monopolar spindle 1 kinase (MPS1; IC50 = ~35 nM). Inhibits the phosphorylation of full-length immunoprecipitated human MPS1 and exhibits minimal activity against a panel of 49 other protein kinases. Has much reduced activity against FAK, JNK1, JNK2, and KIT and does not affect the activity of mitotic-specific phosphoforms of Aurora B and BubR1. Induces a significant reduction in time to complete mitosis (from 90 min to 32 min) and causes abnormal mitosis where over 50% of cells enter anaphase without aligning their chromosomes.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Hewitt, L., et al. 2010. Jour. Cell Biol.190, 25.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service