Skip to Content
Merck
All Photos(1)

Documents

5.30918

Sigma-Aldrich

PTC-124

Synonym(s):

PTC-124, 3-(5-(2-Fluorophenyl)-1,2,4-oxadiazol-3-yl)benzoic acid, Premature Termination Codon Mutations Inhibitor, PTC124, Ataluren

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9FN2O3
CAS Number:
Molecular Weight:
284.24
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C15H9FN2O3/c16-12-7-2-1-6-11(12)14-17-13(18-21-14)9-4-3-5-10(8-9)15(19)20/h1-8H,(H,19,20)

InChI key

OOUGLTULBSNHNF-UHFFFAOYSA-N

General description

A cell-permeable, non-toxic, orally bioavailable oxadiazole compound that is reported to promote read-through of premature, but not normal, translation termination codons and thereby suppress premature termination in protein synthesis caused by nonsense mutations (in ~10 to 100 nM with maximal activity reported at 3 µM). Promotes the suppression of nonsense alleles of the dystrophin gene in primary human muscle cells and in a mouse model of Duchenne muscular dystrophy (mdx). Also reported to rescue striated muscle function in mdx mice with a noticeable reduction in their serum creatine kinase values. Subcutaneous or oral administration of PTC124 to a mice expressing human CFTR-G542X transgene is shown to suppress G542X nonsense mutation and restore hCFTR function and cAMP-stimulated chloride channel activity (60 mg/kg for 3 weeks). Also acts as a potent and reversible inhibitor of purified firefly luciferase (FLuc; IC50 ~ 10 nM) and protects it from tryptic degradation, but does not affect Renilla reniformis luciferase (RLuc).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PTC
Secondary Target
firefly luciferase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

McElroy, S.P., et al. 2013. PLoS Biol.11, e1001593.
Auld, D.S., et al. 2010. Proc. Natl. Acad. Sci. USA.107, 4878.
Auld, D.S., et al. 2009. Proc. Natl. Acad. Sci. USA.106, 3585.
Du, M., et al. 2008. Proc. Natl. Acad. Sci. USA.105, 2064.

Welch, E.M., et al. 2007. Nature.447, 87.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service