Skip to Content
Merck
All Photos(1)

Key Documents

5.30541

Sigma-Aldrich

CFTR Potentiator, VX-770

Synonym(s):

CFTR Potentiator, VX-770, N-(5-Hydroxy-2,4- bis(2-methyl-2-propanyl)phenyl]-4-oxo-1,4-dihydro-3-quinolinecarboxamide, Cystic Fibrosis Transmembrane Conductance Regulator Potentiator, VX 770, Ivacaftor, Kalydeco, N-(5-Hydroxy-2,4-bis(2-methyl-2-propanyl)phenyl]-4-oxo-1,4-dihydro-3-quinolinecarboxamide, Cystic Fibrosis Transmembrane Conductance Regulator Potentiator, VX 770, Ivacaftor, Kalydeco

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N2O3
CAS Number:
Molecular Weight:
392.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Product 5.30541 is not currently sold in your country. Contact Technical Service

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1c2c([c](c(c1)C(=O)Nc3c(cc(c(c3)O)C(C)(C)C)C(C)(C)C)=O)cccc2

InChI

1S/C24H28N2O3/c1-23(2,3)16-11-17(24(4,5)6)20(27)12-19(16)26-22(29)15-13-25-18-10-8-7-9-14(18)21(15)28/h7-13,27H,1-6H3,(H,25,28)(H,26,29)

InChI key

PURKAOJPTOLRMP-UHFFFAOYSA-N

General description

A cell-permeable, orally bioavailable dihydroquinoline-carboxamide compound that acts as a potentiator of Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) by directly binding to WT and G551D mutant forms in an allosteric and reversible manner (~ 10 µM) and increases the open probability to restore lung function. Enhances cAMP/PKA-signaling mediated gating activity of both WT and G551D mutants and its potentiation activity is dependent upon the phosphorylated status of CFTR. Does not compete for binding to the canonical ATP-binding catalytic site; but the rate of flux through CFTR increase by 1.5 to 2 fold when ATP concentrations are increased. Shown to increase Cl- secretion by about 10 fold in cultured human CF bronchial epethelia (HBE) carrying G551D/F508del mutations and increases forskolin-stimulated transepethial current (IT; EC50 = 236 nM) far more effectively than genistein (EC50 = 16 µM). Displays favorable pharmacokinetic properties with t1/2= 12 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
WT and G551D mutant forms of cell surface CFTR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Namkung, W., et al. 2013. Mol. Pharmacol.84, 384.
Eckford, P.D.W., et al. 2012. J. Biol. Chem.287, 36639.

Van Goor, F., et al 2009. Proc. Natl. Acad. Sci. USA106, 18825.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service