Skip to Content
Merck
All Photos(1)

Key Documents

529625

Sigma-Aldrich

Polyphosphoinositide-Binding Peptide, PBP10

A Rhodamine-B-labeled decapeptide derived from the (PIP₂)-binding region in segment-2 of gelsolin.

Synonym(s):

Polyphosphoinositide-Binding Peptide, PBP10, RhoB-GS 160-169, Rhodamine B-QRLFQVKGRR-OH, RhoB-GS160-169, Rhodamine B-QRLFQVKGRR-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H127N24O15
CAS Number:
Molecular Weight:
1713.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pink-red

solubility

DMSO: 2 mg/mL

fluorescence

λex 565 nm
λem 590 nm

shipped in

ambient

storage temp.

−20°C

General description

A Rhodamine-B-labeled decapeptide derived from the (PIP2)-binding region in segment-2 of gelsolin. Acts as a cell-permeable, reversible, and non-toxic agent that transiently disrupts actin filaments, which are essential for cell morphology and cell motility (~1-10 µM). Specifically binds to polyphosphoinositide (PPI) with high affinity and activates platelet without significantly affecting intracellular Ca2+ influx. Undergoes a reduction in fluorescence intensity upon binding to phosphatidylinositol 4,5-bisphosphate (PIP2) and lysophosphatidic acid; the fluorescence is not affected in the presence of phosphatidylserine and phosphatidylcholine. Useful fluorescent probe for the cellular localization and stabilization of PPIs.
A Rhodamine-B-labeled decapeptide derived from the (PIP2)-binding region in segment-2 of gelsolin. Acts as a cell-permeable, reversible, and non-toxic agent that transiently disrupts actin filaments, which are essential for cell morphology and cell motility (~1-10 µM). Specifically binds to polyphosphoinositide (PPI) with high affinity and activates platelets without significantly affecting intracellular Ca2+ influx. Undergoes a reduction in fluorescence intensity upon binding to phosphatidylinositol 4,5-bisphosphate (PIP2) and lysophosphatidic acid; the fluorescence is not affected in the presence of phosphatidylserine and phosphatidylcholine. Useful fluorescent probe for the cellular localization and stabilization of PPIs. Excitation max.: ~565 nm; emission max.: ~590 nm.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Specifically binds to polyphosphoinositide (PPI)
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Rho-B-Gln-Arg-Leu-Phe-Gln-Val-Lys-Gly-Arg-Arg-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bucki, R., et al. 2001. Biochemistry40, 15752
Cunningham, C.C., et al. 2001. J. Biol. Chem.276, 43390.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service