Skip to Content
Merck
All Photos(1)

Documents

521231

Sigma-Aldrich

PDGFR Tyrosine Kinase Inhibitor II

The PDGFR Tyrosine Kinase Inhibitor II, also referenced under CAS 249762-74-1, controls the biological activity of PDGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PDGFR Tyrosine Kinase Inhibitor II, (5-Butanoate-1H-2-indolyl)(1H-2-indolyl)-methanone, [2-(1H-2-Indolylcarbonyl)-1H-5-indolyl]butanoate, D-65476

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N2O3
CAS Number:
Molecular Weight:
346.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable bis(1H-2-indolyl)-1-methanone compound that acts as a highly selective, ATP-competitive and reversible inhibitor of platelet-derived growth factor (PDGF) receptor tyrosine kinase (IC50 = 1.1 M in Swiss 3T3 cells for PDGFR). Also potently inhibits Flt3 (Fms-like tyrosine kinase 3) activity (IC50 = 6.2 M for hPDGFβ-R-mFlt3 and 50 nM in EOL-1 cells). Sensitizes 32D cells to radiation-induced apoptosis and inhibits proliferation of TEL-Flt3-transfected BA/F3 cells (IC50<300 nM). May also serve as a prodrug form of the Platelet Derived Growth Factor Receptor Tyrosine Kinase Inhibitor I in cells.
Prodrug form of Platelet Derived Growth Factor Receptor Tyrosine Kinase Inhibitor I (Cat. No. 521230) that acts as a highly selective, cell-permeable, ATP-competitive, and reversible inhibitor of platelet-derived growth factor receptor (PDGFR) tyrosine kinase (IC50 = 1.1 µM, Swiss 3T3 cells). Also potently inhibits Fms-like tyrosine kinase 3 (Flt3) activity (IC50 = 6.2 µM for PDGFRβ-mFlt3 and 50 nM in EOL-1 cells). Reported to sensitize 32D cells to radiation-induced apoptosis. Also shown to inhibit proliferation of TEL-Flt3-transfected BA/F3 cells (IC50 >300 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDGFR in swiss 3T3 cells
Product competes with ATP.
Reversible: yes
Target IC50: 1.1 µM inhibiting PDGFR in swiss 3T3 cells; 6.2 µ M for hPDGFβ-R-mFlt3 and 50 nM in EOL-1 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mahboobi, S., et al. 2002. J. Med. Chem.45, 1002.
Teller, S., et al. 2002. Leukemia16, 1528.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service