Skip to Content
Merck
All Photos(1)

Key Documents

5.08507

Sigma-Aldrich

Isradipine

Synonym(s):

Isradipine, L-type voltage-gated Ca2+ channel blocker, Isradipine, PN 200-110

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€134.00

€134.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€134.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O5
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€134.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mM
ethanol: 50 mM

storage temp.

2-8°C

SMILES string

N1C(=C(C(C(=C1C)C(=O)OC)c2c3n[o]nc3ccc2)C(=O)OC(C)C)C

InChI

1S/C19H21N3O5/c1-9(2)26-19(24)15-11(4)20-10(3)14(18(23)25-5)16(15)12-7-6-8-13-17(12)22-27-21-13/h6-9,16,20H,1-5H3

InChI key

HMJIYCCIJYRONP-UHFFFAOYSA-N

General description

A water soluble voltage gated calcium channel gating inhibitor of the dihydropyridine class. In human ion channels, Isradipine has gating inhibition on Cav1.4 channels (1x10E-6 M) and Cav1.3 (3x10E-8) and in mouse ion channels a pIC50 of 6.7. Often prescribed for hypertension (trade names DynaCirc, Prescal), recent animal models suggest a use in Parkinson′s Disease.
A water soluble voltage gated calcium channel gating inhibitor of the dihydropyridine class. In human ion channels, Isradipine has gating inhibition on Cav1.4 channels (1x10E-6 M) and Cav1.3 (3x10E-8) and in mouse ion channels a pIC50 of 6.7. Often prescribed for hypertension (trade names DynaCirc, Prescal), recent animal models suggest a use in Parkinson′s Disease.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
CaV

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chan, C. S. et al. 2007. Nature.447, 1081.
Baumann, L. et al. 2004. Invest. Ophthalmol. Vis. Sci.45, 708.
Koschak, A. et al. 2003. J. Neurosci.23, 6041.
Koschak, A. et al. 2001. J. Biol Chem276, 22100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service