Skip to Content
Merck
All Photos(1)

Documents

506190

Sigma-Aldrich

p97 ATPase Activity Inhibitor, DBeQ

The p97 ATPase Activity Inhibitor, DBeQ controls the biological activity of p97 ATPase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

p97 ATPase Activity Inhibitor, DBeQ, CID676352, CID16472035, N²,N⁴-Dibenzylquinazolinine-2,4-diamine, KUC105555N, Valosin-containing Protein Inhibitor I, VCP Inhibitor I, ERAD Inhibitor I, p97 Inhibitor I, Valosin-containing Protein Inhibitor I, VCP Inhibitor I, CID676352, CID16472035, N²,N⁴-Dibenzylquinazolinine-2,4-diamine, KUC105555N, ERAD Inhibitor I, p97 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4
Molecular Weight:
340.42
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 2.5 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable quinazolinamine compound that inhibits the AAA (ATPase associated with diverse cellular activities) ATPase p97 (IC50 = 1.6 µM against WT or C522A p97) in a reversible and ATP-competitive (Ki = 3.2 µM) manner, while being at least 50-fold less potent against AAA ATPase NSF (N-methylmaleimide-sensitive factor) or the ATP-dependent chymotrypsin-like activity of proteasome 20S, and exhibiting no significant activity against a panel of ~170 kinases. Shown to effectively suppress p97-dependent cellular processes, including ERAD (ER-associated degradation) reporter TCRα-GFP degradation (92.59% vs. 33.33% degradation in 2 h, respectively, with DMSO or 10 µM DBeQ in HEK293 cultures), autophagic degradation of LC3-II (LC3-II/LC3-I = 0.04 vs. 1.0, respectively, after 3 h DMSO or 10 µM DBeQ treatment in HeLa cultures), UFD (ubiquitin fusion degradation) pathway-mediated UbG76V-GFP reporter degradation (IC50 = 2.3 µM in HeLa cells), but not the degradation of p97-independent proteasome substrates ODC (ornithine decarboxylase) and ODD (oxygen-dependent degradation domain of HIF-1α) luciferase fusion reporters (IC50 = 45 and 56 µM, respectively, in HeLa cells). Also demonstrated to display selective antiproliferative activity against RPMI8226, HeLa, and HEK293 over non-cancerous human fetal lung fibroblasts MRC5 (GI50 = 1.2, 3.1, 4.0, and 6.6 µM, respectively) by inducing cellular DEVD-like (caspase-3/7), but not VEID-like (caspase-6), activity (by 4- to 6-fold in 4 h in HeLa cells) via a yet unknown mechanism.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chou, T.F., et al. 2011. Proc. Natl. Acad. Sci. USA108, 4834.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service