5.05855
Adenovirus Proteinase Inhibitor
powder, ≥90% (HPLC)
Synonym(s):
Adenovirus Proteinase Inhibitor, NSC 37249, 2-(Dicyclohexylamino)-N-(4-(4-((2-(dicyclohexylamino)acetyl)amino)phenyl)sulfonylphenyl)acetamide, N,Nʹ-(Sulfonyldi-4,1-phenylene)bis(2-(dicyclohexylamino)acetamide, AVP Inhibitor, AVP Inhibitor, 2-(Dicyclohexylamino)-N-(4-(4-((2-(dicyclohexylamino)acetyl)amino)phenyl)sulfonylphenyl)acetamide, N,Nʹ-(Sulfonyldi-4,1-phenylene)bis(2-(dicyclohexylamino)acetamide
Select a Size
€263.00
Select a Size
About This Item
€263.00
Recommended Products
Product Name
Adenovirus Proteinase Inhibitor, NSC 37249,
Assay
≥90% (HPLC)
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
5% TFA in ethanol: 5 mg/mL
DMSO: insoluble
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(c4ccc(cc4)NC(=O)CN(C6CCCCC6)C5CCCCC5)c1ccc(cc1)NC(=O)CN(C3CCCCC3)C2CCCCC2
InChI
1S/C40H58N4O4S/c45-39(29-43(33-13-5-1-6-14-33)34-15-7-2-8-16-34)41-31-21-25-37(26-22-31)49(47,48)38-27-23-32(24-28-38)42-40(46)30-44(35-17-9-3-10-18-35)36-19-11-4-12-20-36/h21-28,33-36H,1-20,29-30H2,(H,41,45)(H,42,46)
InChI key
JQYJJPSTWWRURQ-UHFFFAOYSA-N
General description
Biochem/physiol Actions
AVP co-factor pVIc (GVQSLKRRRCF) N-terminal binding pocket
AVP-pVIc substrate-binding site
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service