Skip to Content
Merck
All Photos(1)

Key Documents

5.05762

Sigma-Aldrich

(+/-)-Clopidogrel Hydrogensulfate

Synonym(s):

(+/-)-Clopidogrel Hydrogensulfate, Purinergic P2Y12 Receptor Antagonist, (+/-)-Clopidogrel, Plavix

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€237.00

€237.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2S · xH2SO4
CAS Number:
Molecular Weight:
321.82 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€237.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])O.[s]1c2c(cc1)C[N+H](CC2)C(c3c(cccc3)Cl)C(=O)OC

InChI

1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)

InChI key

FDEODCTUSIWGLK-UHFFFAOYSA-N

General description

A thienopyridine compound that acts as a specific irreversible inhibitor of purinergic P2Y12 receptors. Blocks ADP-induced platelet aggregation and serves as an anti-thrombotic agent. Shown to reduce vascular inflammation and angiotensin II induced-abdominal aortic aneurysm progression. Clopidogrel is a prodrug that requires activation by CYP enzyme system to produce its pharmacologically active metabolite.

Biochem/physiol Actions

Primary Target
P2Y₁₂
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, O., et al. 2012. PLoS One.7, e51707.

Savi, P., et al. 2005. Semin. Thromb. Hemost.31, 174.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service