Skip to Content
Merck
All Photos(1)

Key Documents

5.05479

Sigma-Aldrich

SIRT1/2/3 pan Inhibitor

Synonym(s):

SIRT1/2/3 pan Inhibitor, 4-(4-(2-Pivalamidoethyl)piperidin-1-yl)thieno[3,2-d]pyrimidine-6-carboxamide, pan SIRT1/2/3 Inhibitor, pan SIRT1/2/3 Inhibitor, 4-(4-(2-Pivalamidoethyl)piperidin-1-yl)thieno[3,2-d]pyrimidine-6-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€251.00

€251.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€251.00

About This Item

Empirical Formula (Hill Notation):
C19H27N5O2S
CAS Number:
Molecular Weight:
389.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€251.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A thienopyrimidinylcarboxamide that acts as a highly potent, reversible pan sirtuin (SIRT) inhibitor (IC50 = 15, 10, and 33 nM for SIRT1, 2, and 3, respectively). Shown to bind to the SIRT catalytic site and occupy both the nicotinamide C-pocket and acetyl lysine substrate channel. Exhibits high selectivity over a panel of protein kinases, nuclelar receptors, GPCRs, and ion-channels (IC50 >10 µM). Poorly affects hERG and cytochrome P450s (>50 µM). Exhibits desirable stability in microsomal preparations (human CLint = 15.8 µL/min/mg, mouse CLint = 12.7 µL/min/mg) and high solubility (297 µM), and low LogD (2.73).

Biochem/physiol Actions

Primary Target
SIRT1/2/3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Disch, J.S., et al. 2013. J. Med. Chem.57, 1275.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service