Skip to Content
Merck
All Photos(1)

Key Documents

5.05051

Sigma-Aldrich

2,3 cis-Piperidine dicarboxylic acid

Synonym(s):

2,3 cis-Piperidine dicarboxylic acid, Ionotropic Glutamate Receptor Antagonist, PDA, cis PDA, cis-Piperidine-2,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€147.00

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€147.00

About This Item

Empirical Formula (Hill Notation):
C7H11NO4
CAS Number:
Molecular Weight:
173.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mM

storage temp.

2-8°C

SMILES string

N1C(C(CCC1)C(=O)O)C(=O)O

InChI

1S/C7H11NO4/c9-6(10)4-2-1-3-8-5(4)7(11)12/h4-5,8H,1-3H2,(H,9,10)(H,11,12)

InChI key

PTLWNCBCBZZBJI-UHFFFAOYSA-N

General description

A non-specific antagonist of NMDA, AMPA and kainate ionotropic receptors and a partial agonist for NMDA receptors. Used in blocking general excitatory synaptic transmissions.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tong, G., et al. 1994. Neuron13, 1195.
Leach, J., et al. 1986. Eur. J. Pharmacol.121, 173.
Watkins, C., et al. 1981. Annu. Rev. Pharmacol. Toxicol.21, 165.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service