Skip to Content
Merck
All Photos(1)

Key Documents

5.04845

Sigma-Aldrich

Ro 25-6981 Maleate Salt

Synonym(s):

Ro 25-6981 Maleate Salt, [R-(R*,S*)]-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO2 · xC4H4O4
CAS Number:
Molecular Weight:
339.47 (free base basis)
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.009 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

H2O: 25 mM

storage temp.

2-8°C

InChI

1S/C22H29NO2/c1-17(22(25)20-7-9-21(24)10-8-20)16-23-13-11-19(12-14-23)15-18-5-3-2-4-6-18/h2-10,17,19,22,24-25H,11-16H2,1H3/t17-,22+/m0/s1

InChI key

WVZSEUPGUDIELE-HTAPYJJXSA-N

General description

A potent, activity-dependent antagonist highly selective for GluN1/NR2B subtype NMDA receptors (IC50 = 0.009 and 52 µM for NR2B and NR2A subtype receptors respectively). Commonly used for studying brain functions and disorders involving neurotransmissions mediated by GluN1/NR2B subtype NMDA receptors (learning and memory, neuroplasticity, neurotoxicity, neuropathic pain, addiction, and neurogenesis enhancement).

Biochem/physiol Actions

Primary Target
GluN1/NR2B receptors

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Qi, Y., et al. 2013. Cereb. Cortex.23, 932.
Soloviova, A., et al. 2012. Exp. Biol. & Med.153, 764.
Solovyeva, A., et al. 2012. Neurosci. & Behav. Physiol.42, 456.
Chazot, L., et al. 2004. Curr. Med. Chem.11, 389.
Lynch, R., et al. 2001. Eur. J. Pharmacol.416, 185.
Fischer, G., et al. 1997. J. Pharmacol. Exp. Ther.283, 1285.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service