Skip to Content
Merck
All Photos(1)

Documents

494461-M

Millipore

Octyl β-D-1-thioglucopyranoside

ULTROL® Grade, ≥98% (HPLC), non-ionic detergent to isolate membrane proteins from E. coli without inactivation

Synonym(s):

n-Octyl-β-D-thioglucopyranoside, ULTROL® Grade, OSGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O5S
CAS Number:
Molecular Weight:
308.43
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.28

Quality Level

100
300

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

CMC

9 mM

solubility

water: 25%

storage temp.

10-30°C

InChI

1S/C14H28O5S/c1-2-3-4-5-6-7-8-20-14-13(18)12(17)11(16)10(9-15)19-14/h10-18H,2-9H2,1H3

InChI key

CGVLVOOFCGWBCS-UHFFFAOYSA-N

General description

Non-ionic detergent that has been used to isolate membrane proteins from E. coli without inactivation. Stable in aqueous solution and easily removed by dialysis. Highly purified and intended for use as a detergent to solubilize membrane-bound proteins in their native state (CMC: 9 mM).
Non-ionic detergent that has been used to isolate membrane proteins from E. coli without inactivation. Stable in aqueous solution and easily removed by dialysis. Highly purified and intended for use as a detergent to solubilize membrane-bound proteins in their native state.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 4 months at 4°C.

Other Notes

Saito, S., and Tsuchiya, T. 1984. Biochem. J.222, 829.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service