Skip to Content
Merck

4.80940

SAFC

Ethanol 20 % (v/v) with 150 mMol/L sodium chloride solution

EMPROVE® EXPERT

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

950 L
€10,292.00

€10,292.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
950 L
€10,292.00

About This Item

UNSPSC Code:
41103206

€10,292.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

product line

EMPROVE® EXPERT

Quality Level

transition temp

flash point 35 °C

density

0.970 g/cm3 at 20 °C

format

mixture

storage temp.

2-30°C

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service