Skip to Content
Merck
All Photos(1)

Documents

475872

Sigma-Aldrich

MnTMPyP

Cell-permeable superoxide dismutase (SOD) mimetic.

Synonym(s):

MnTMPyP, Mn(III)tetrakis(1-methyl-4-pyridyl)porphyrin, Tetratosylate, Hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H65MnN8O13S4
Molecular Weight:
1433.53
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

black

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Cell-permeable superoxide dismutase (SOD) mimetic. Catalyzes the dismutation of O2– • (rate constant ~4 x 107 M-1 s-1) even in the presence of excess EDTA. Can substitute for SOD in mutants of E. coli lacking this enzyme. MnTMPyP acts in vivo as an NADPH/GSH:O2 oxidoreductase rather than as an SOD mimetic. Also an efficient peroxynitrite reductase when redox-coupled with biological antioxidants.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Izbicka, E., et al. 1999. Cancer Res.59, 639.
Lee, J., et al. 1998. J. Am. Chem. Soc. 120, 6053.
Gardner, P.R., et al. 1996. Arch. Biochem. Biophys. 325, 20.
Liochev, S.I., and Fridovich, I. 1995. Arch. Biochem. Biophys. 321, 271.
Faulkner, K.M., et al. 1994. J. Biol. Chem. 269, 23471.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service