Skip to Content
Merck
All Photos(1)

Key Documents

444282

Sigma-Aldrich

MMP/TACE Substrate, FS-6

The widely used MMP substrate FS-1 is modified by an N-terminal lysine elongation to produce a broad-spectrum MMP and TACE substrate that displays improved water solubility and substrate sensitivities.

Synonym(s):

MMP/TACE Substrate, FS-6, Mca-KPLG~L-Dpa-AR-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€327.00

€327.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€327.00

About This Item

Empirical Formula (Hill Notation):
C55H80N16O16
Molecular Weight:
1221.32
UNSPSC Code:
12352200
NACRES:
NA.77

€327.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

The widely used MMP substrate FS-1 (Cat. No. 03-32-5032) is modified by an N-terminal lysine elongation to produce a broad-spectrum MMP and TACE substrate that displays improved water solubility and substrate sensitivities. Exhibits 2- to 9-fold higher specificity constant (kcat/Km) towards MMP-1, -8, -13, -14 when compared with FS-1 and ~2-fold higher kcat/Km towards TACE when compared with TACE Substrate II and III (Cat. Nos. 616402 and 616403, respectively). It has been used successfully in detecting T98G glioma cell surface-associated MMP activity and is shown to be resistant to proteolysis by human leukocyte elastase, trypsin, and ADAM-10.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Broad-spectrum MMP and TACE substrate
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Mca-Lys-Pro-Leu-Gly~Leu-Dpa-Ala-Arg-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Neumann, U., et al. 2004. Anal. Biochem.328, 166.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service