Skip to Content
Merck
All Photos(1)

Key Documents

407241

Sigma-Aldrich

Insulin Degrading Enzyme, His•Tag®, Rat, Recombinant, S. frugiperda

Synonym(s):

Insulysin, IDE

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€484.00

€484.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€484.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.25

€484.00


Please contact Customer Service for Availability

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥3 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, rat insulin degrading enzyme fused to a His•Tag sequence and expressed in S. frugiperda insect cells. Insulin degrading enzyme (Insulysin) is a metalloprotease that degrades insulin and a variety of other peptides, including glucagon, insulin-like growth factors I and II, atrial natriuretic peptide, TGF-α, and amyloid peptides. Reported to be inversely correlated with brain Aβ peptide levels in insulysin-deficient mice compared to wild-type mice.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of the enzyme that will catalyze the hydrolysis of 1 µmol iodinated insulin per h at 37°C, pH 7.3.

Physical form

In 100 mM potassium phosphate buffer, 20% glycerol, pH 7.3.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Hersh, L.B. 2003. Curr. Pharm. Des.9, 449.
Miller, B.C., et al. 2003. Proc. Natl. Acad. Sci. USA100, 6221.
Song, E.S., et al. 2003. J. Biol. Chem.278, 49789.
Mukherjee, A., and Hersh, L.B. 2002. J. Alzheimer′s Dis.4, 341.
Song, E.S., et al. 2001. J. Biol. Chem.276, 1152.
Becker, A.B., and Roth, R.A. 1995. Methods Enzymol.248, 693.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service