Skip to Content
Merck
All Photos(1)

Key Documents

251635

Sigma-Aldrich

Daminozide

Synonym(s):

Daminozide, Histone Lysine Demethylase Inhibitor XII, LSD1 Inhibitor VI, N,N-Dimethylamino-succinamic acid, Succinic acid-2,2-dimethylhydrazide, 4-(2,2-Dimethylhydrazinyl)-4-oxobutanoic acid, BHC100 Inhibitor VI, KDM2A Inhibitor, KIAA1718 Inhibitor VI, LSD Inhibitor VI, PHF8 Inhibitor, JHDM Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O3
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (NMR)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)

InChI key

NOQGZXFMHARMLW-UHFFFAOYSA-N

General description

A dimethylamino substituted succinamate that, in addition to its well known use as a plant growth regulator, also acts as an inhibitor against histone lysine demthylases KDM2A, LSD1 (KDM7A; KIAA1718), and PHF8 (IC50 = 1.5, 2.1, and 0.55 µM, respectively), but not KDM3A, KDM4E, KDM5C, KDM6B, or three hydroxylases (IC50 >100 µM). Substrate-competition and/or active site Iron chelating likely accounts for the inhibitory activity against the FAD-dependent LSD1, while 2-oxoglutarate (2-OG) competition (Ki = 1.97 µM) appears as the major mode of action against KDM2A.
A dimethylamino substituted succinamic acid compound that, in addition to its well known use as a plant growth regulator, also acts as an inhibitor against histone lysine demthylases KDM2A, LSD1 (KDM7A; KIAA1718), and PHF8 (IC50 = 1.5, 2.1, and 0.55 µM, respectively), but not KDM3A, KDM4E, KDM5C, KDM6B, (IC50 >100 µM) or three hydroxylases (FIH, PHD2, and BBOX1; IC50 >100 µM). Although kinetic studies indicate 2-oxoglutarate (2-OG) competition (Ki = 1.97 µM) as the major mode of action against KDM2A, substrate-competition and/or active site Iron chelation more likely accounts for the compound′s inhibitory activity against the FAD-dependent LSD1.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Rose, N.R., et al. 2012. J. Med. Chem.55, 6639.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service